Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Ethyl-2-methylpyridine, 97%

Chemical Identifiers

Chemical Name or Material 5-Ethyl-2-methylpyridine
CAS 104-90-5
Molecular Formula C8H11N
Formula Weight 121.18
MDL Number MFCD00006344
UN Number UN2300
Beilstein 109269
Merck Index 14,3844
Synonym 5-Ethyl-2-picoline; 2-Methyl-5-ethylpyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0690122

  SDS
alfa aesar™
L0690122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0690136

  SDS
alfa aesar™
L0690136
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 121.18
Chemical Name or Material 5-Ethyl-2-methylpyridine
Molecular Formula C8H11N
CAS 104-90-5
Health Hazard 1 Danger
TSCA TSCA
Assay 97%
MDL Number MFCD00006344
UN Number UN2300
Merck Index 14,3844
Synonym 5-Ethyl-2-picoline; 2-Methyl-5-ethylpyridine
Health Hazard 2 GHS H Statement
H311-H331-H314-H318-H227-H302
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 178° to 179°C
Melting Point −71°C
Quantity 100g
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: 2-METHYL-5-ETHYLPYRIDINE
RTECS Number TJ6825000
Beilstein 109269
Flash Point 66°C (150°F)
Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H311-H331-H314-H318-H227-H302
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.