Learn More
5-Ethyl-2-methylpyridine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0690136
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5-Ethyl-2-methylpyridine | |
-71°C | |
178°C to 179°C | |
C8H11N | |
MFCD00006344 | |
UN2300 | |
14,3844 | |
NTSLROIKFLNUIJ-UHFFFAOYSA-N | |
5-ethyl-2-methylpyridine | |
7728 | |
97% |
104-90-5 | |
0.919 | |
66°C (150°F) | |
1.497 | |
500 g | |
109269 | |
aldehydine, 5-ethyl-2-picoline, 2-methyl-5-ethylpyridine, aldehydkollidin, aldehydecollidine, pyridine, 5-ethyl-2-methyl, 2,5-aldehydine, 2-picoline, 5-ethyl, 3-ethyl-6-methylpyridine, 6-methyl-3-ethylpyridine | |
CCC1=CC=C(C)N=C1 | |
121.18 | |
121.18 |
Chemical Identifiers
104-90-5 | |
121.18 | |
NTSLROIKFLNUIJ-UHFFFAOYSA-N | |
7728 | |
CCC1=CC=C(C)N=C1 |
C8H11N | |
MFCD00006344 | |
aldehydine, 5-ethyl-2-picoline, 2-methyl-5-ethylpyridine, aldehydkollidin, aldehydecollidine, pyridine, 5-ethyl-2-methyl, 2,5-aldehydine, 2-picoline, 5-ethyl, 3-ethyl-6-methylpyridine, 6-methyl-3-ethylpyridine | |
5-ethyl-2-methylpyridine |
Safety and Handling
GHS H Statement
H311-H331-H314-H318-H227-H302
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
P210-P233-P240-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P330-P363-P374-P501c
H227-H302-H311+H331-H314
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: 2-METHYL-5-ETHYLPYRIDINE
EINECSNumber : 203-250-0
RTECSNumber : TJ6825000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only