Please login to your online account to display your discounted pricing

5-Ethyl-2-furaldehyde, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-Ethyl-2-furaldehyde
CAS 23074-10-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7 H8 O2
Formula Weight 124.14
MDL Number MFCD00030605
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC336550050

  SDS
thermo fisher scientific
336550050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 124.14
Quantity 5g
Color Clear colorless to dark yellow
Density 1.0500g/mL
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C7 H8 O2
MDL Number MFCD00030605
CAS Min % 97.5
Chemical Name or Material 5-Ethyl-2-furaldehyde
Packaging Glass bottle
Refractive Index 1.5215 to 1.5235 (20°C, 589nm)
CAS 23074-10-4