Please login to your online account to display your discounted pricing

5-Ethyl-2-furaldehyde, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  336550050

 View more versions of this product

Catalog No. AC336550050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 124.14
Physical Form Liquid
Quantity 5g
Color Clear colorless to dark yellow
Density 1.0500g/mL
CAS Max % 100.0
Assay Percent Range 98%
Infrared Spectrum Authentic
CAS Min % 97.5
Molecular Formula C7 H8 O2
MDL Number MFCD00030605
Chemical Name or Material 5-Ethyl-2-furaldehyde
CAS 23074-10-4
Packaging Glass bottle
Refractive Index 1.5215 to 1.5235 (20°C, 589nm)

Specifications

Chemical Name or Material 5-Ethyl-2-furaldehyde
CAS 23074-10-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7 H8 O2
Formula Weight 124.14
MDL Number MFCD00030605