missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Ethyl-2,3-dimethylpyrazine, TCI America™

Manufacturer:  TCI America E08285G

 View more versions of this product

Catalog No. E08285G



Specifications

Specifications

5-Ethyl-2,3-dimethylpyrazine
Yellow
15707-34-3
MFCD04038059
CCC1=CN=C(C(=N1)C)C
136.198
136.20
≥98.0% (GC,T)
76°C
5g
C8H12N2
CIBKSMZEVHTQLG-UHFFFAOYSA-N
5-ethyl-2,3-dimethylpyrazine
27460
Clear Liquid at 20°C

Chemical Identifiers

15707-34-3
136.198
CIBKSMZEVHTQLG-UHFFFAOYSA-N
5-ethyl-2,3-dimethylpyrazine
C8H12N2
MFCD04038059
27460
CCC1=CN=C(C(=N1)C)C
Safety and Handling

Safety and Handling

TSCA : No

Documents