missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Ethoxycarbonyl-2-fluorobenzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H53134MD

 View more versions of this product

Catalog No. AAH53134MD



Specifications

Specifications

5-Ethoxycarbonyl-2-fluorobenzeneboronic acid
250mg
C9H10BFO4
5-ethoxycarbonyl-2-fluorophenyl boronic acid, 3-borono-4-fluorobenzoic acid ethyl ester, 5-ethoxycarbonyl-2-fluorobenzeneboronic acid, 2-fluoro-5-ethoxycarbonylphenylboronic acid, 5-ethoxycarbonyl-2-fluorophenylboronic acid, benzoic acid,3-borono-4-fluoro-, 1-ethyl ester 9ci, 5-ethoxycarbonyl-2-fluorophenyl boronic acid, acmc-209qlk, 5-ethoxycarbonyl-2-fluorophenylboronic acid contains varying amounts of anhydride
B(C1=C(C=CC(=C1)C(=O)OCC)F)(O)O
211.983
211.98
97°C to 99°C
874219-60-0
MFCD08235094
QXIIYFKTSGJTNE-UHFFFAOYSA-N
(5-ethoxycarbonyl-2-fluorophenyl)boronic acid
44717604
97%

Chemical Identifiers

874219-60-0
211.983
QXIIYFKTSGJTNE-UHFFFAOYSA-N
44717604
B(C1=C(C=CC(=C1)C(=O)OCC)F)(O)O
C9H10BFO4
MFCD08235094
5-ethoxycarbonyl-2-fluorophenyl boronic acid, 3-borono-4-fluorobenzoic acid ethyl ester, 5-ethoxycarbonyl-2-fluorobenzeneboronic acid, 2-fluoro-5-ethoxycarbonylphenylboronic acid, 5-ethoxycarbonyl-2-fluorophenylboronic acid, benzoic acid,3-borono-4-fluoro-, 1-ethyl ester 9ci, 5-ethoxycarbonyl-2-fluorophenyl boronic acid, acmc-209qlk, 5-ethoxycarbonyl-2-fluorophenylboronic acid contains varying amounts of anhydride
(5-ethoxycarbonyl-2-fluorophenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents