Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Ethoxy-2-mercaptobenzimidazole, 97%

$40.65 - $130.22

Chemical Identifiers

Chemical Name or Material 5-Ethoxy-2-mercaptobenzimidazole
CAS 55489-15-1
Molecular Formula C9H10N2OS
Formula Weight 194.26
MDL Number MFCD01326487
Beilstein 4310013
Synonym 5-Ethoxybenzimidazole-2-thiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1778106

 
alfa aesar™
L1778106
5g Each for $40.65

AAL1778114

 
alfa aesar™
L1778114
25g Each for $130.22
Description & Specifications

Specifications

Formula Weight 194.26
Chemical Name or Material 5-Ethoxy-2-mercaptobenzimidazole
CAS 55489-15-1
Health Hazard 1 Warning
Assay 97%
Molecular Formula C9H10N2OS
MDL Number MFCD01326487
Beilstein 4310013
Synonym 5-Ethoxybenzimidazole-2-thiol
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 246° to 250°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.