Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Dimethylamino-2-methyl-3-pentyn-2-ol, 98%

$30.45 - $30.45

Chemical Identifiers

Chemical Name or Material 5-Dimethylamino-2-methyl-3-pentyn-2-ol
CAS 25400-83-3
Molecular Formula C8H15NO
Formula Weight 141.21
MDL Number MFCD00041575
Beilstein 1750347
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1225603

  SDS
alfa aesar™
L1225603
1g Each for $30.45
Description & Specifications

Specifications

Formula Weight 141.21
Chemical Name or Material 5-Dimethylamino-2-methyl-3-pentyn-2-ol
Assay 98%
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
CAS 25400-83-3
MDL Number MFCD00041575
Health Hazard 1 Warning
Boiling Point 91° to 92°C (1mmHg)
Molecular Formula C8H15NO
Beilstein 1750347
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.