missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Dimethylamino-2-methyl-3-pentyn-2-ol, 98%

Manufacturer:  Alfa Aesar™ L1225606

 View more versions of this product

Catalog No. AAL1225606



Specifications

Specifications

5-Dimethylamino-2-methyl-3-pentyn-2-ol
5g
C8H15NO
1750347
VBARYRPUABJYGW-UHFFFAOYSA-N
5-(dimethylamino)-2-methylpent-3-yn-2-ol
180775
98%
91°C to 92°C (1mmHg)
25400-83-3
MFCD00041575
5-dimethylamino-2-methyl-3-pentyn-2-ol, 5-dimethylamino-2-methylpent-3-yn-2-ol, 5-dimethylamino-2-methyl-3-pentyn-2-ol, 3-pentyn-2-ol, 5-dimethylamino-2-methyl, acmc-1cdlj, 5-dimethylamino-2-methyl-pent-3-yn-2-ol, 3-pentyn-2-ol,5-dimethylamino-2-methyl
CC(C)(C#CCN(C)C)O
141.214
141.21

Chemical Identifiers

25400-83-3
141.214
VBARYRPUABJYGW-UHFFFAOYSA-N
180775
CC(C)(C#CCN(C)C)O
C8H15NO
MFCD00041575
5-dimethylamino-2-methyl-3-pentyn-2-ol, 5-dimethylamino-2-methylpent-3-yn-2-ol, 5-dimethylamino-2-methyl-3-pentyn-2-ol, 3-pentyn-2-ol, 5-dimethylamino-2-methyl, acmc-1cdlj, 5-dimethylamino-2-methyl-pent-3-yn-2-ol, 3-pentyn-2-ol,5-dimethylamino-2-methyl
5-(dimethylamino)-2-methylpent-3-yn-2-ol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Documents