missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Difluoromethoxy-2-mercaptobenzimidazole, 97%

$21.45 - $84.19

Chemical Identifiers

CAS 97963-62-7
Molecular Formula C8H6F2N2OS
Molecular Weight (g/mol) 216.206
MDL Number MFCD00467504
InChI Key HJMVPNAZPFZXCP-UHFFFAOYSA-N
Synonym 5-difluoromethoxy-2-mercapto-1h-benzimidazole, 5-difluoromethoxy-1h-benzo d imidazole-2-thiol, 5-difluoromethoxy-2-mercapto-1h-benzimidazole, 5-difluoromethoxy-1h-benzimidazole-2-thiol, unii-39p59c89nv, 5-difluoromethoxy-2-benzimidazolethiol, 5-difluoromethoxy-2-mercaptobenzimidazole, 5-difluoromethoxy-2-thio-benzimidazole, 5-difluoromethoxy-1h-1,3-benzodiazole-2-thiol, 5-difluoromethoxy-1,3-dihydrobenzimidazole-2-thione
PubChem CID 5064774
IUPAC Name 5-(difluoromethoxy)-1,3-dihydrobenzimidazole-2-thione
SMILES C1=CC2=C(C=C1OC(F)F)NC(=S)N2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6037503 View Documents Alfa Aesar™
H6037503
1g Each for $21.45
 
AAH6037506 View Documents Alfa Aesar™
H6037506
5g Each for $84.19
 
Specifications

Chemical Identifiers

97963-62-7
216.206
HJMVPNAZPFZXCP-UHFFFAOYSA-N
5064774
C1=CC2=C(C=C1OC(F)F)NC(=S)N2
C8H6F2N2OS
MFCD00467504
5-difluoromethoxy-2-mercapto-1h-benzimidazole, 5-difluoromethoxy-1h-benzo d imidazole-2-thiol, 5-difluoromethoxy-2-mercapto-1h-benzimidazole, 5-difluoromethoxy-1h-benzimidazole-2-thiol, unii-39p59c89nv, 5-difluoromethoxy-2-benzimidazolethiol, 5-difluoromethoxy-2-mercaptobenzimidazole, 5-difluoromethoxy-2-thio-benzimidazole, 5-difluoromethoxy-1h-1,3-benzodiazole-2-thiol, 5-difluoromethoxy-1,3-dihydrobenzimidazole-2-thione
5-(difluoromethoxy)-1,3-dihydrobenzimidazole-2-thione

Specifications

239°C to 243°C
97963-62-7
MFCD00467504
HJMVPNAZPFZXCP-UHFFFAOYSA-N
5-(difluoromethoxy)-1,3-dihydrobenzimidazole-2-thione
5064774
97%
1g
C8H6F2N2OS
5-difluoromethoxy-2-mercapto-1h-benzimidazole, 5-difluoromethoxy-1h-benzo d imidazole-2-thiol, 5-difluoromethoxy-2-mercapto-1h-benzimidazole, 5-difluoromethoxy-1h-benzimidazole-2-thiol, unii-39p59c89nv, 5-difluoromethoxy-2-benzimidazolethiol, 5-difluoromethoxy-2-mercaptobenzimidazole, 5-difluoromethoxy-2-thio-benzimidazole, 5-difluoromethoxy-1h-1,3-benzodiazole-2-thiol, 5-difluoromethoxy-1,3-dihydrobenzimidazole-2-thione
C1=CC2=C(C=C1OC(F)F)NC(=S)N2
216.206
216.21
5-Difluoromethoxy-2-mercaptobenzimidazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : −30° to −10°C