missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5'-Deoxy-5-fluorocytidine 98.0+%, TCI America™
$76.43 - $228.38
Chemical Identifiers
| CAS | 66335-38-4 |
|---|---|
| Molecular Formula | C9H12FN3O4 |
| Molecular Weight (g/mol) | 245.21 |
| MDL Number | MFCD07369278 |
| InChI Key | YSNABXSEHNLERR-ZIYNGMLESA-N |
| PubChem CID | 10037499 |
| ChEBI | CHEBI:80627 |
| IUPAC Name | 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-methyloxolan-2-yl]-5-fluoro-1,2-dihydropyrimidin-2-one |
| SMILES | C[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=C(F)C(N)=NC1=O |
Chemical Identifiers
| 66335-38-4 | |
| 245.21 | |
| YSNABXSEHNLERR-ZIYNGMLESA-N | |
| CHEBI:80627 | |
| C[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=C(F)C(N)=NC1=O |
| C9H12FN3O4 | |
| MFCD07369278 | |
| 10037499 | |
| 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-methyloxolan-2-yl]-5-fluoro-1,2-dihydropyrimidin-2-one |
Specifications
| 66335-38-4 | |
| White | |
| MFCD07369278 | |
| YSNABXSEHNLERR-ZIYNGMLESA-N | |
| 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-methyloxolan-2-yl]-5-fluoro-1,2-dihydropyrimidin-2-one | |
| 10037499 | |
| 245.21 | |
| Crystalline Powder |
| 204°C | |
| C9H12FN3O4 | |
| 1 g | |
| C[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=C(F)C(N)=NC1=O | |
| 245.21 | |
| CHEBI:80627 | |
| ≥98.0% (HPLC,T) | |
| 5′-Deoxy-5-fluorocytidine |
Safety and Handling
TSCA : No