Learn More
5-Decyne, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1305018
Description
5-Decyne is used as APIs and intermediates. It is also used for chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Decyne is used as APIs and intermediates. It is also used for chemical research.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
5-Decyne | |
0.77 | |
54°C (129°F) | |
C10H18 | |
MFCD00027277 | |
UN3295 | |
5-decyne, dibutylacetylene, 1,2-dibutylacetylene, acmc-209ey1, 5-decyne 8ci 9ci, 3,3?-diheptyloxacarbocyanine iodide | |
JWBQJUFCNOLNNC-UHFFFAOYSA-N | |
dec-5-yne | |
16030 | |
98% |
1942-46-7 | |
177°C to 178°C | |
Petrol-like | |
1.433 | |
50 g | |
1697840 | |
Not miscible in water. | |
CCCCC#CCCCC | |
138.254 | |
138.26 |
Chemical Identifiers
1942-46-7 | |
138.254 | |
JWBQJUFCNOLNNC-UHFFFAOYSA-N | |
16030 | |
CCCCC#CCCCC |
C10H18 | |
MFCD00027277 | |
5-decyne, dibutylacetylene, 1,2-dibutylacetylene, acmc-209ey1, 5-decyne 8ci 9ci, 3,3?-diheptyloxacarbocyanine iodide | |
dec-5-yne |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only