missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Decyn-1-ol, 97%

Manufacturer:  Alfa Aesar™ H5340014

 View more versions of this product

Catalog No. AAH5340014



Description

Description

Applications
5-Decyn-1-ol is used in the synthesis of multi-substituted cyclopropanes. It involved in manufacture of fine chemicals, having applications in electroplating and perfumery industry.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

5-Decyn-1-ol
25g
C10H18O
MFCD00464353
RNTNGQCWAKHVKO-UHFFFAOYSA-N
dec-5-yn-1-ol
109957
97%
238°C
68274-97-5
CH3(CH2)3C≡C(CH2)4OH
5-decyn-1-ol, 5-decyn-1-ol, 5-decyn-1-ol,
CCCCC#CCCCCO
154.253
154.25

Chemical Identifiers

68274-97-5
154.253
RNTNGQCWAKHVKO-UHFFFAOYSA-N
109957
CCCCC#CCCCCO
C10H18O
MFCD00464353
5-decyn-1-ol, 5-decyn-1-ol, 5-decyn-1-ol,
dec-5-yn-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents