Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Cyclopropyl-1H-pyrazole-3-carboxylic acid, 94%

$112.68 - $372.50

Chemical Identifiers

Chemical Name or Material 5-Cyclopropyl-1H-pyrazole-3-carboxylic acid
CAS 401629-04-7
Molecular Formula C7H8N2O2
Formula Weight 152.15
MDL Number MFCD04967194
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5030603

 
alfa aesar™
H5030603
1g Each for $372.50

AAH50306MD

 
alfa aesar™
H50306MD
250mg Each for $112.68
Description & Specifications

Specifications

Formula Weight 152.15
Chemical Name or Material 5-Cyclopropyl-1H-pyrazole-3-carboxylic acid
Assay 94%
Molecular Formula C7H8N2O2
MDL Number MFCD04967194
CAS 401629-04-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈175°C
Quantity 1g

Applications
5-Cyclopropyl-1H-pyrazole-3-carboxylic acid is used as organic chemical synthesis intermediate.

Notes
Air sensitive. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.