missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-[Cyclohexyl(methyl)carbamoyl]-2-fluorobenzeneboronic acid, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H54567MD

 View more versions of this product

Catalog No. AAH54567MD



Specifications

Specifications

5-[Cyclohexyl(methyl)carbamoyl]-2-fluorobenzeneboronic acid
1449131-79-6
MFCD20040224
PIAHGOURQDHKCB-UHFFFAOYSA-N
[5-[cyclohexyl(methyl)carbamoyl]-2-fluorophenyl]boronic acid
73995878
97%
250mg
C14H19BFNO3
5-cyclohexyl methyl carbamoyl-2-fluorobenzeneboronic acid
B(C1=C(C=CC(=C1)C(=O)N(C)C2CCCCC2)F)(O)O
279.118
279.11

Chemical Identifiers

1449131-79-6
279.118
PIAHGOURQDHKCB-UHFFFAOYSA-N
73995878
B(C1=C(C=CC(=C1)C(=O)N(C)C2CCCCC2)F)(O)O
C14H19BFNO3
MFCD20040224
5-cyclohexyl methyl carbamoyl-2-fluorobenzeneboronic acid
[5-[cyclohexyl(methyl)carbamoyl]-2-fluorophenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents