Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Cyclobutyl-5-phenylhydantoin, 97+%

$73.53 - $292.31

Chemical Identifiers

Chemical Name or Material 5-Cyclobutyl-5-phenylhydantoin
CAS 125650-44-4
Molecular Formula C13H14N2O2
Formula Weight 230.27
MDL Number MFCD00022395
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1611503

  SDS
alfa aesar™
A1611503
1g Each for $73.53

AAA1611506

  SDS
alfa aesar™
A1611506
5g Each for $292.31
Description & Specifications

Specifications

Formula Weight 230.27
Chemical Name or Material 5-Cyclobutyl-5-phenylhydantoin
Assay ≥97%
Molecular Formula C13H14N2O2
MDL Number MFCD00022395
CAS 125650-44-4
Melting Point 234° to 235°C
Quantity 1g