missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Cyanomethyl-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
Chemical Identifiers
CAS | 1637543-22-6 |
---|---|
Molecular Formula | C14H17BFNO2 |
Molecular Weight (g/mol) | 261.103 |
InChI Key | XPHLMDUDMYOYHY-UHFFFAOYSA-N |
Synonym | 5-Cyanomethyl-2-fluorophenylboronic acid pinacol ester |
PubChem CID | 100913436 |
IUPAC Name | 2-[4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)CC#N)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH62223MD
|
Thermo Scientific Chemicals
H62223MD |
250mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1637543-22-6 | |
261.103 | |
5-Cyanomethyl-2-fluorophenylboronic acid pinacol ester | |
2-[4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile |
C14H17BFNO2 | |
XPHLMDUDMYOYHY-UHFFFAOYSA-N | |
100913436 | |
B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)CC#N)F |
Specifications
1637543-22-6 | |
250mg | |
XPHLMDUDMYOYHY-UHFFFAOYSA-N | |
2-[4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile | |
100913436 | |
96% |
C14H17BFNO2 | |
5-Cyanomethyl-2-fluorophenylboronic acid pinacol ester | |
B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)CC#N)F | |
261.103 | |
261.1 | |
5-Cyanomethyl-2-fluorobenzeneboronic acid pinacol ester |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
TSCA : No
Recommended Storage : Ambient temperatures