missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Cyano-3-(3-oxocyclopentyl)indole, 95%

Manufacturer:  Alfa Aesar™ H63625MD

 View more versions of this product

Catalog No. AAH63625MD



Specifications

Specifications

5-Cyano-3-(3-oxocyclopentyl)indole
675831-64-8
UN3439
PCCIEBVCPXEIFS-UHFFFAOYSA-N
3-(3-oxocyclopentyl)-1H-indole-5-carbonitrile
22139459
Solid
250mg
C14H12N2O
3-3-oxocyclopentyl-1h-indole-5-carbonitrile, 3-3-oxocyclopentyl-5-cyano-1h-indole, 3-3-oxo-cyclopentyl-1h-indole-5-carbonitrile, 1h-indole-5-carbonitrile, 3-3-oxocyclopentyl
C1CC(=O)CC1C2=CNC3=C2C=C(C=C3)C#N
224.263
224.26
95%

Chemical Identifiers

675831-64-8
224.263
3-3-oxocyclopentyl-1h-indole-5-carbonitrile, 3-3-oxocyclopentyl-5-cyano-1h-indole, 3-3-oxo-cyclopentyl-1h-indole-5-carbonitrile, 1h-indole-5-carbonitrile, 3-3-oxocyclopentyl
3-(3-oxocyclopentyl)-1H-indole-5-carbonitrile
C14H12N2O
PCCIEBVCPXEIFS-UHFFFAOYSA-N
22139459
C1CC(=O)CC1C2=CNC3=C2C=C(C=C3)C#N
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

SDS Documents