missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Cyano-3-(1-methyl-1,2,3,6-tetrahydro-4-pyridyl)indole, 97%

Manufacturer:  Alfa Aesar™ H63834MD

 View more versions of this product

Catalog No. AAH63834MD



Specifications

Specifications

5-Cyano-3-(1-methyl-1,2,3,6-tetrahydro-4-pyridyl)indole
250mg
C15H15N3
UN3439
3-1-methyl-1,2,3,6-tetrahydro-4-pyridinyl-1h-indole-5-carbonitrile, 3-1-methyl-1,2,3,6-tetrahydropyridin-4-yl-1h-indole-5-carbonitrile, 3-1-methyl-3,6-dihydro-2h-pyridin-4-yl-1h-indole-5-carbonitrile, methyltetrahydropyridinylindolecarbonitrile, 3-1-methyl-4-1,2,5,6-tetrahydropyridyl indole-5-carbonitrile, 5-cyano-3-1-methyl-1,2,3,6-tetrahydropyridin-4-yl-1h-indole, 3-1-methyl-1,2,3,6-tetrahydro-4-pyridyl-1h-indole-5-carbonitrile, 3-1-methyl-1,2,3,6-tetrahydro-pyridin-4-yl-1h-indole-5-carbonitrile, 3-1-methyl-1,2,3,6-tetrahydropyridin-4-yl-1h-indole-5-carbonitrile
CN1CCC(=CC1)C2=CNC3=C2C=C(C=C3)C#N
237.306
237.3
97%
White to Yellow
116480-60-5
MFCD08276951
11193296
HNOKHIURIJOQKM-UHFFFAOYSA-N
3-(1-methyl-3,6-dihydro-2H-pyridin-4-yl)-1H-indole-5-carbonitrile
14973216
Solid

Chemical Identifiers

116480-60-5
237.306
HNOKHIURIJOQKM-UHFFFAOYSA-N
14973216
CN1CCC(=CC1)C2=CNC3=C2C=C(C=C3)C#N
C15H15N3
MFCD08276951
3-1-methyl-1,2,3,6-tetrahydro-4-pyridinyl-1h-indole-5-carbonitrile, 3-1-methyl-1,2,3,6-tetrahydropyridin-4-yl-1h-indole-5-carbonitrile, 3-1-methyl-3,6-dihydro-2h-pyridin-4-yl-1h-indole-5-carbonitrile, methyltetrahydropyridinylindolecarbonitrile, 3-1-methyl-4-1,2,5,6-tetrahydropyridyl indole-5-carbonitrile, 5-cyano-3-1-methyl-1,2,3,6-tetrahydropyridin-4-yl-1h-indole, 3-1-methyl-1,2,3,6-tetrahydro-4-pyridyl-1h-indole-5-carbonitrile, 3-1-methyl-1,2,3,6-tetrahydro-pyridin-4-yl-1h-indole-5-carbonitrile, 3-1-methyl-1,2,3,6-tetrahydropyridin-4-yl-1h-indole-5-carbonitrile
3-(1-methyl-3,6-dihydro-2H-pyridin-4-yl)-1H-indole-5-carbonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

SDS Documents