Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Cyano-2-(trifluoromethoxy)benzeneboronic acid, 97%

Chemical Identifiers

Chemical Name or Material 5-Cyano-2-(trifluoromethoxy)benzeneboronic acid
CAS 1072946-64-5
Formula Weight 230.94
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6117703

 
alfa aesar™
H6117703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH61177MD

 
alfa aesar™
H61177MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 230.94
Chemical Name or Material 5-Cyano-2-(trifluoromethoxy)benzeneboronic acid
CAS 1072946-64-5
Physical Form Solid
Assay 97%
Quantity 1g