Please login to your online account to display your discounted pricing

5-Cyano-2-methylpyridine, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 5-Cyano-2-methylpyridine
Assay Percent Range 97%
Molecular Formula C7H6N2
Formula Weight 118.14
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC452420010

  SDS
thermo fisher scientific
AC452420010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC452420050

  SDS
thermo fisher scientific
AC452420050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 118.14
Quantity 1g
Chemical Name or Material 5-Cyano-2-methylpyridine
Assay Percent Range 97%
Molecular Formula C7H6N2
CAS 3222-48-8