Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Cyano-2-hydroxybenzeneboronic acid, 96%

Chemical Identifiers

Chemical Name or Material 5-Cyano-2-hydroxybenzeneboronic acid
CAS 1256355-57-3
Formula Weight 162.94
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH61915MD

 
alfa aesar™
H61915MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 162.94
Chemical Name or Material 5-Cyano-2-hydroxybenzeneboronic acid
Physical Form Solid
Assay 96%
CAS 1256355-57-3
Quantity 250mg