missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Cyano-2-fluorophenylboronic acid, 97%, ACROS Organics™

Chemical Identifiers

CAS 468718-30-1
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C7H5BFNO2
Formula Weight 164.93
MDL Number MFCD03095147
Chemical Name or Material 5-Cyano-2-fluorophenylboronic acid
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC430230010 SDS Thermo Fisher Scientific
AC430230010
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC430230050 SDS Thermo Fisher Scientific
AC430230050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

468718-30-1
100.0
C7H5BFNO2
MFCD03095147
≥96.0%
96.0
97%
164.93
5-Cyano-2-fluorophenylboronic acid

Specifications

Crystalline Powder
1g
96.0
MFCD03095147
468718-30-1
Authentic
≥96.0%
164.93
White
100.0
C7H5BFNO2
97%
5-Cyano-2-fluorophenylboronic acid

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.