missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5α-Cholestane, 98+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC165600050

 View more versions of this product

Catalog No. AC165600050



Description

Description

Specifications

Specifications

5α-Cholestane
Glass bottle
+ 26.5
White
5g
98.0
98% min. (GC)
MFCD00066412
28,29,30-trinorlanostane, 5-alpha-cholestane, 5?-cholestane, 5alpha-cholestane, 5alpha-cholestane 8ci, alpha-cholestane, cholestane, cholestane van, unii-u260hwn305
XIIAYQZJNBULGD-XWLABEFZSA-N
(5R,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthrene
2723895
372.67
98+%
Authentic
+ 26.50 (20.00°C c=2,CHCl3)
250°C (1.0mmHg)
78°C to 81°C
481-21-0
100.0
C27H48
15, 2206
Solubility in water: insoluble in water. Other solubilities: freely soluble in chloroform (50mg/mL),ether and,slightly soluble in absolute alcohol
CC(C)CCCC(C)C1CCC2C1(CCC3C2CCC4C3(CCCC4)C)C
372.67
CHEBI:35515
Crystalline Powder

Chemical Identifiers

481-21-0
372.67
XIIAYQZJNBULGD-XWLABEFZSA-N
2723895
(5R,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthrene
C27H48
MFCD00066412
28,29,30-trinorlanostane, 5-alpha-cholestane, 5?-cholestane, 5alpha-cholestane, 5alpha-cholestane 8ci, alpha-cholestane, cholestane, cholestane van, unii-u260hwn305
CHEBI:35515
CC(C)CCCC(C)C1CCC2C1(CCC3C2CCC4C3(CCCC4)C)C
Safety and Handling

Safety and Handling

EINECSNumber : 207-562-8

SDS Documents