Please login to your online account to display your discounted pricing

5α -Cholest-8(14)-en-3β-ol-15-one, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 5α -cholest-8(14)-en-3β-ol-15-one
CAS 50673-97-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C27H44O2
Formula Weight 400.64
MDL Number MFCD09065035
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB01709EA

  SDS
fisher scientific company
SB01709EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 400.64
Quantity 10g
Assay 97%
CAS Min % 97.0
Molecular Formula C27H44O2
Packaging Amber glass bottle
CAS 50673-97-7
Chemical Name or Material 5α -cholest-8(14)-en-3β-ol-15-one
Assay Percent Range 97%
MDL Number MFCD09065035