Please login to your online account to display your discounted pricing

5α -Cholest-8(14)-en-3β-ol-15-one, 97%, Maybridge


Quantity 10g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  SB01709EA

 View more versions of this product

Catalog No. SB01709EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 400.64
Quantity 10g
Assay 97%
CAS Min % 97.0
Packaging Amber glass bottle
Molecular Formula C27H44O2
CAS 50673-97-7
Chemical Name or Material 5α -cholest-8(14)-en-3β-ol-15-one
Assay Percent Range 97%
MDL Number MFCD09065035

Specifications

Chemical Name or Material 5α -cholest-8(14)-en-3β-ol-15-one
CAS 50673-97-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C27H44O2
Formula Weight 400.64
MDL Number MFCD09065035