missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chlorovanillin, 96%

$113.76 - $113.76

Chemical Identifiers

CAS 19463-48-0
Molecular Formula C8H7ClO3
Formula Weight 186.59
MDL Number MFCD00016982
Chemical Name or Material 5-Chlorovanillin
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2555406 SDS Alfa Aesar™
B2555406
5g Each for $113.76
Specifications Description & Specifications

Chemical Identifiers

19463-48-0
186.59
5-Chlorovanillin
C8H7ClO3
MFCD00016982

Specifications

162° to 166°C
19463-48-0
186.59
5-Chlorovanillin
5g
C8H7ClO3
MFCD00016982
96%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning