Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chlorovaleronitrile, 97%

Chemical Identifiers

Chemical Name or Material 5-Chlorovaleronitrile
CAS 6280-87-1
Molecular Formula Cl(CH2)4CN
Formula Weight 117.58
MDL Number MFCD00001977
UN Number UN3276
Beilstein 1742079
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0141309

  SDS
alfa aesar™
L0141309
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0141318

  SDS
alfa aesar™
L0141318
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 117.58
Molecular Formula Cl(CH2)4CN
MDL Number MFCD00001977
CAS 6280-87-1
Chemical Name or Material 5-Chlorovaleronitrile
Assay 97%
UN Number UN3276
Beilstein 1742079
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
Flash Point 98°C (208°F)
Boiling Point 92° to 93°C (10mmHg)
Quantity 10g
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.