missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chlorouracil, 98%

Manufacturer:  Alfa Aesar™ A1108406

 View more versions of this product

Catalog No. AAA1108406



Specifications

Specifications

5-Chlorouracil
5g
98%
MFCD00006019
5-Chloro-2,4-dihydroxypyrimidine
C1=C(C(=O)NC(=O)N1)Cl
146.53
CHEBI:60762
>300°C
1820-81-1
C4H3ClN2O2
127173
ZFTBZKVVGZNMJR-UHFFFAOYSA-N
5-chloro-1H-pyrimidine-2,4-dione
15758
146.53

Chemical Identifiers

1820-81-1
146.53
ZFTBZKVVGZNMJR-UHFFFAOYSA-N
15758
5-chloro-1H-pyrimidine-2,4-dione
C4H3ClN2O2
MFCD00006019
5-Chloro-2,4-dihydroxypyrimidine
CHEBI:60762
C1=C(C(=O)NC(=O)N1)Cl
Safety and Handling

Safety and Handling

RTECSNumber : YQ9410000

SDS Documents