missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Chlorouracil, 98%, Thermo Scientific Chemicals
$127.14 - $404.00
Chemical Identifiers
CAS | 1820-81-1 |
---|---|
Molecular Formula | C4H3ClN2O2 |
Molecular Weight (g/mol) | 146.53 |
MDL Number | MFCD00006019 |
InChI Key | ZFTBZKVVGZNMJR-UHFFFAOYSA-N |
Synonym | 5-chlorouracil, 2,4 1h,3h-pyrimidinedione, 5-chloro, uracil, 5-chloro, 5-chloro-2,4-dihydroxypyrimidine, 5-chloropyrimidine-2,4 1h,3h-dione, unii-7lq4v03rny, 7lq4v03rny, 5-chloro-1,3-dihydropyrimidine-2,4-dione, 5-chloropyrimidine-2,4 1h,3h-dione 5-chlorouracil, zlchem 428 |
PubChem CID | 15758 |
ChEBI | CHEBI:60762 |
SMILES | ClC1=CNC(=O)NC1=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1108406
|
Thermo Scientific Chemicals
A1108406 |
5 g |
Each for $127.14
|
|
|||||
AAA1108414
|
Thermo Scientific Chemicals
A1108414 |
25 g |
Each for $404.00
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1820-81-1 | |
146.53 | |
ZFTBZKVVGZNMJR-UHFFFAOYSA-N | |
15758 | |
ClC1=CNC(=O)NC1=O |
C4H3ClN2O2 | |
MFCD00006019 | |
5-chlorouracil, 2,4 1h,3h-pyrimidinedione, 5-chloro, uracil, 5-chloro, 5-chloro-2,4-dihydroxypyrimidine, 5-chloropyrimidine-2,4 1h,3h-dione, unii-7lq4v03rny, 7lq4v03rny, 5-chloro-1,3-dihydropyrimidine-2,4-dione, 5-chloropyrimidine-2,4 1h,3h-dione 5-chlorouracil, zlchem 428 | |
CHEBI:60762 |
Specifications
1820-81-1 | |
C4H3ClN2O2 | |
5 g | |
5-chlorouracil, 2,4 1h,3h-pyrimidinedione, 5-chloro, uracil, 5-chloro, 5-chloro-2,4-dihydroxypyrimidine, 5-chloropyrimidine-2,4 1h,3h-dione, unii-7lq4v03rny, 7lq4v03rny, 5-chloro-1,3-dihydropyrimidine-2,4-dione, 5-chloropyrimidine-2,4 1h,3h-dione 5-chlorouracil, zlchem 428 | |
ClC1=CNC(=O)NC1=O | |
146.53 | |
CHEBI:60762 | |
98% |
>300°C | |
MFCD00006019 | |
127173 | |
ZFTBZKVVGZNMJR-UHFFFAOYSA-N | |
5-chloro-1,2,3,4-tetrahydropyrimidine-2,4-dione | |
15758 | |
146.53 | |
5-Chlorouracil |
Safety and Handling
EINECSNumber : 217-339-7
RTECSNumber : YQ9410000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only