missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Chlorotryptamine Hydrochloride, TCI America™

$85.56 - $304.03

Chemical Identifiers

CAS 942-26-7
Molecular Formula C10H12Cl2N2
Molecular Weight (g/mol) 231.12
MDL Number MFCD00051990
InChI Key PBANXRNIXGEHPZ-UHFFFAOYSA-N
Synonym 5-chlorotryptamine hydrochloride, 2-5-chloro-1h-indol-3-yl ethanamine hydrochloride, 5-chlorotryptamine hcl, 5-chloro-1h-indole-3-ethylamine hydrochloride, 5-chloro-1h-indole-3-ethylamine monohydrochloride, 5-chlorotryptamine monohydrochloride, 2-5-chloro-1h-indol-3-yl ethylamine hydrochloride, 5-chloro-1h-indole-3-ethylamine hcl, 2-5-chloro-1h-indol-3-yl ethan-1-amine hydrochloride, 5-chloro-3-2-aminoethyl indole hydrochloride
PubChem CID 2827494
IUPAC Name 2-(5-chloro-1H-indol-3-yl)ethanamine;hydrochloride
SMILES C1=CC2=C(C=C1Cl)C(=CN2)CCN.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C11455G View Documents TCI America
C11455G
5g Each for $85.56
 
C114525G View Documents TCI America
C114525G
25g Each for $304.03
 
Specifications

Chemical Identifiers

942-26-7
231.12
PBANXRNIXGEHPZ-UHFFFAOYSA-N
2827494
C1=CC2=C(C=C1Cl)C(=CN2)CCN.Cl
C10H12Cl2N2
MFCD00051990
5-chlorotryptamine hydrochloride, 2-5-chloro-1h-indol-3-yl ethanamine hydrochloride, 5-chlorotryptamine hcl, 5-chloro-1h-indole-3-ethylamine hydrochloride, 5-chloro-1h-indole-3-ethylamine monohydrochloride, 5-chlorotryptamine monohydrochloride, 2-5-chloro-1h-indol-3-yl ethylamine hydrochloride, 5-chloro-1h-indole-3-ethylamine hcl, 2-5-chloro-1h-indol-3-yl ethan-1-amine hydrochloride, 5-chloro-3-2-aminoethyl indole hydrochloride
2-(5-chloro-1H-indol-3-yl)ethanamine;hydrochloride

Specifications

Undesignated
942-26-7
MFCD00051990
PBANXRNIXGEHPZ-UHFFFAOYSA-N
2-(5-chloro-1H-indol-3-yl)ethanamine;hydrochloride
2827494
Crystal-Powder at 20°C
5-Chlorotryptamine Hydrochloride
5g
C10H12Cl2N2
5-chlorotryptamine hydrochloride, 2-5-chloro-1h-indol-3-yl ethanamine hydrochloride, 5-chlorotryptamine hcl, 5-chloro-1h-indole-3-ethylamine hydrochloride, 5-chloro-1h-indole-3-ethylamine monohydrochloride, 5-chlorotryptamine monohydrochloride, 2-5-chloro-1h-indol-3-yl ethylamine hydrochloride, 5-chloro-1h-indole-3-ethylamine hcl, 2-5-chloro-1h-indol-3-yl ethan-1-amine hydrochloride, 5-chloro-3-2-aminoethyl indole hydrochloride
C1=CC2=C(C=C1Cl)C(=CN2)CCN.Cl
231.12
231.12
≥98.0% (N)
Safety and Handling

Safety and Handling

TSCA : No