Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chlorotryptamine hydrochloride, 98%

$230.54 - $3,646.36

Chemical Identifiers

Chemical Name or Material 5-Chlorotryptamine hydrochloride
CAS 942-26-7
Molecular Formula C10H11ClN2·HCl
Formula Weight 231.13
MDL Number MFCD00051990
Beilstein 3718105
Synonym 2-(5-Chloro-3-indolyl)ethylamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2480503

  SDS
alfa aesar™
B2480503
1g Each for $230.54

AAB2480506

  SDS
alfa aesar™
B2480506
5g Each for $913.09

AAB2480514

  SDS
alfa aesar™
B2480514
25g Each for $3,646.36
Description & Specifications

Specifications

Formula Weight 231.13
Chemical Name or Material 5-Chlorotryptamine hydrochloride
Assay 98%
Molecular Formula C10H11ClN2·HCl
MDL Number MFCD00051990
CAS 942-26-7
Beilstein 3718105
Synonym 2-(5-Chloro-3-indolyl)ethylamine hydrochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈290°C (dec.)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.