missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-chlorothiophenesulphonyl chloride, 96%, ACROS Organics™

Chemical Identifiers

CAS 2766-74-7
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula C4 H2 Cl2 O2 S2
Formula Weight 217.1
MDL Number MFCD00051667
Chemical Name or Material 5-Chlorothiophenesulphonyl chloride
Assay ≥95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC342260050 SDS Thermo Fisher Scientific
34226-0050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

2766-74-7
100.0
C4 H2 Cl2 O2 S2
MFCD00051667
≥95%
95.0
96%
217.1
5-Chlorothiophenesulphonyl chloride

Specifications

Crystalline Low Melting Solid or Liquid
5g
95.0
96%
C4 H2 Cl2 O2 S2
MFCD00051667
Authentic
≥95%
217.1
White to yellow
100.0
Product may darken during storage
2766-74-7
Glass bottle
5-Chlorothiophenesulphonyl chloride

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger