Learn More
Alfa Aesar™ 5-Chlorothiophene-2-carboxylic acid, 98%
Manufacturer: Alfa Aesar™ A1520922
Specifications
5-Chlorothiophene-2-carboxylic acid | |
100g | |
C5H3ClO2S | |
118361 | |
QZLSBOVWPHXCLT-UHFFFAOYSA-N | |
5-chlorothiophene-2-carboxylic acid | |
95048 | |
98% |
149°C to 152°C | |
24065-33-6 | |
MFCD00041426 | |
5-chloro-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 5-chloro, 2-chloro-5-carboxythiophene, 5-chloro-2-thiophene carboxylic acid, pubchem7380, maybridge3_003717, acmc-209g8i, rivaroxaban related compound, 5-18-06-00177 beilstein handbook reference, ksc204m2j | |
C1=C(SC(=C1)Cl)C(=O)O | |
162.587 | |
162.6 |
Chemical Identifiers
24065-33-6 | |
162.587 | |
QZLSBOVWPHXCLT-UHFFFAOYSA-N | |
95048 | |
C1=C(SC(=C1)Cl)C(=O)O |
C5H3ClO2S | |
MFCD00041426 | |
5-chloro-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 5-chloro, 2-chloro-5-carboxythiophene, 5-chloro-2-thiophene carboxylic acid, pubchem7380, maybridge3_003717, acmc-209g8i, rivaroxaban related compound, 5-18-06-00177 beilstein handbook reference, ksc204m2j | |
5-chlorothiophene-2-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
RTECSNumber : XM8400000