Learn More
5-Chlorothiophene-2-boronic acid, 97+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 359010050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
5-Chlorothiophene-2-boronic acid | |
97+% | |
C4H4BClO2S | |
5g | |
Solubility in water: insoluble | |
OB(O)C1=CC=C(Cl)S1 | |
162.39 | |
162.4 |
162607-18-3 | |
Glass bottle | |
MFCD00236030 | |
5-chlorothiophene-2-boronic acid, 5-chloro-2-thienylboronic acid, 5-chlorothiophen-2-yl boronic acid, 5-chloro-2-thiopheneboronic acid, 5-chlorothiophen-2-boronic acid, 5-chlorothiophen-2-yl boranediol, 5-chlorothiphene 2-boronic acid, 5-chloro-2-thienyl boronic acid, boronic acid, 5-chloro-2-thienyl, 5-chlorothiophene-2-boronicacid | |
JFCLNCVCDFUJPO-UHFFFAOYSA-N | |
(5-chlorothiophen-2-yl)boronic acid | |
2734324 | |
97+% |
Chemical Identifiers
162607-18-3 | |
162.39 | |
JFCLNCVCDFUJPO-UHFFFAOYSA-N | |
2734324 | |
OB(O)C1=CC=C(Cl)S1 |
C4H4BClO2S | |
MFCD00236030 | |
5-chlorothiophene-2-boronic acid, 5-chloro-2-thienylboronic acid, 5-chlorothiophen-2-yl boronic acid, 5-chloro-2-thiopheneboronic acid, 5-chlorothiophen-2-boronic acid, 5-chlorothiophen-2-yl boranediol, 5-chlorothiphene 2-boronic acid, 5-chloro-2-thienyl boronic acid, boronic acid, 5-chloro-2-thienyl, 5-chlorothiophene-2-boronicacid | |
(5-chlorothiophen-2-yl)boronic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
RUO â Research Use Only