missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Chlorothiophene-2-boronic acid, 97+%, ACROS Organics™

Chemical Identifiers

CAS 162607-18-3
Molecular Formula C4H4BClO2S
Formula Weight 162.4
MDL Number MFCD00236030
Synonym 5-Chloro-2-thienylboronic acid
Chemical Name or Material 5-Chlorothiophene-2-boronic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC359010050 SDS Thermo Fisher Scientific
AC359010050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC359010010 SDS Thermo Fisher Scientific
AC359010010
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

162607-18-3
162.4
5-Chloro-2-thienylboronic acid
C4H4BClO2S
MFCD00236030
5-Chlorothiophene-2-boronic acid

Specifications

5g
Glass bottle
162.4
5-Chloro-2-thienylboronic acid
162607-18-3
C4H4BClO2S
MFCD00236030
5-Chlorothiophene-2-boronic acid