missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chlorothiophene-2-boronic acid, 97%

$41.14 - $456.26

Chemical Identifiers

CAS 162607-18-3
Molecular Formula C4H4BClO2S
Formula Weight 162.4
MDL Number MFCD00236030
Beilstein 8478804
Synonym 5-Chloro-2-thienylboronic acid
Chemical Name or Material 5-Chlorothiophene-2-boronic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2319314 SDS Alfa Aesar™
B2319314
25g Each for $456.26
AAB2319306 SDS Alfa Aesar™
B2319306
5g Each for $118.62
AAB2319303 SDS Alfa Aesar™
B2319303
1g Each for $41.14
Specifications Description & Specifications

Chemical Identifiers

162607-18-3
162.4
8478804
5-Chlorothiophene-2-boronic acid
C4H4BClO2S
MFCD00236030
5-Chloro-2-thienylboronic acid

Specifications

≈136° to 140°C
162607-18-3
162.4
8478804
5-Chlorothiophene-2-boronic acid
25g
C4H4BClO2S
MFCD00236030
5-Chloro-2-thienylboronic acid
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C