Learn More
5-(chlorosulfonyl)-2-fluorobenzoic acid, 97%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC438010010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
5-(chlorosulfonyl)-2-fluorobenzoic acid | |
96.0 | |
White | |
33.82 to 36.64% C, 1.62 to 1.76% H | |
Authentic | |
Glass bottle | |
1g | |
C1=CC(=C(C=C1S(=O)(=O)Cl)C(=O)O)F | |
238.62 | |
238.62 | |
97% |
37098-75-2 | |
100.0 | |
111.0°C to 113.0°C | |
97% | |
C7H4ClFO4S | |
2-fluoro-5-chlorosulfonyl benzoic acid, 2-fluoro-5-chlorosulfonylbenzoic acid, 5-chlorosulfonyl-2-fluoro-benzoic acid, 5-chlorosulfonyl-2-fluorobenzoic acid, 5-chlorosulfonyl-2-fluorobenzoicacid, 5-chlorosulphonyl-2-fluorobenzoic acid, acmc-1afaw, benzoic acid, 5-chlorosulfonyl-2-fluoro, ksc577o0r | |
CISMZCVQNVNWJW-UHFFFAOYSA-N | |
5-chlorosulfonyl-2-fluorobenzoic acid | |
608984 | |
Crystalline Powder |
Chemical Identifiers
37098-75-2 | |
238.62 | |
2-fluoro-5-chlorosulfonyl benzoic acid, 2-fluoro-5-chlorosulfonylbenzoic acid, 5-chlorosulfonyl-2-fluoro-benzoic acid, 5-chlorosulfonyl-2-fluorobenzoic acid, 5-chlorosulfonyl-2-fluorobenzoicacid, 5-chlorosulphonyl-2-fluorobenzoic acid, acmc-1afaw, benzoic acid, 5-chlorosulfonyl-2-fluoro, ksc577o0r | |
5-chlorosulfonyl-2-fluorobenzoic acid |
C7H4ClFO4S | |
CISMZCVQNVNWJW-UHFFFAOYSA-N | |
608984 | |
C1=CC(=C(C=C1S(=O)(=O)Cl)C(=O)O)F |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth.
Do
GHS Signal Word: Danger
RUO – Research Use Only