missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(chlorosulfonyl)-2-fluorobenzoic acid, 97%, ACROS Organics™

Chemical Identifiers

CAS 37098-75-2
Molecular Formula C7H4ClFO4S
Molecular Weight (g/mol) 238.62
InChI Key CISMZCVQNVNWJW-UHFFFAOYSA-N
Synonym 2-fluoro-5-chlorosulfonyl benzoic acid, 2-fluoro-5-chlorosulfonylbenzoic acid, 5-chlorosulfonyl-2-fluoro-benzoic acid, 5-chlorosulfonyl-2-fluorobenzoic acid, 5-chlorosulfonyl-2-fluorobenzoicacid, 5-chlorosulphonyl-2-fluorobenzoic acid, acmc-1afaw, benzoic acid, 5-chlorosulfonyl-2-fluoro, ksc577o0r
PubChem CID 608984
IUPAC Name 5-chlorosulfonyl-2-fluorobenzoic acid
SMILES C1=CC(=C(C=C1S(=O)(=O)Cl)C(=O)O)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC438010010
View Documents
Thermo Fisher Scientific
AC438010010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC438010050
View Documents
Thermo Fisher Scientific
AC438010050
5g Glass Bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

37098-75-2
238.62
2-fluoro-5-chlorosulfonyl benzoic acid, 2-fluoro-5-chlorosulfonylbenzoic acid, 5-chlorosulfonyl-2-fluoro-benzoic acid, 5-chlorosulfonyl-2-fluorobenzoic acid, 5-chlorosulfonyl-2-fluorobenzoicacid, 5-chlorosulphonyl-2-fluorobenzoic acid, acmc-1afaw, benzoic acid, 5-chlorosulfonyl-2-fluoro, ksc577o0r
5-chlorosulfonyl-2-fluorobenzoic acid
C7H4ClFO4S
CISMZCVQNVNWJW-UHFFFAOYSA-N
608984
C1=CC(=C(C=C1S(=O)(=O)Cl)C(=O)O)F

Specifications

33.82 to 36.64% C, 1.62 to 1.76% H
Glass bottle
111.0°C to 113.0°C
37098-75-2
100.0
C7H4ClFO4S
CISMZCVQNVNWJW-UHFFFAOYSA-N
5-chlorosulfonyl-2-fluorobenzoic acid
608984
Crystalline Powder
5-(chlorosulfonyl)-2-fluorobenzoic acid
Authentic
White
1g
96.0
97%
2-fluoro-5-chlorosulfonyl benzoic acid, 2-fluoro-5-chlorosulfonylbenzoic acid, 5-chlorosulfonyl-2-fluoro-benzoic acid, 5-chlorosulfonyl-2-fluorobenzoic acid, 5-chlorosulfonyl-2-fluorobenzoicacid, 5-chlorosulphonyl-2-fluorobenzoic acid, acmc-1afaw, benzoic acid, 5-chlorosulfonyl-2-fluoro, ksc577o0r
C1=CC(=C(C=C1S(=O)(=O)Cl)C(=O)O)F
238.62
238.62
97%
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth.
Do

GHS Signal Word: Danger