Learn More
5-(chlorosulfonyl)-2-fluorobenzoic acid, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC438010050
Specifications
5-(chlorosulfonyl)-2-fluorobenzoic acid | |
Authentic | |
White | |
5g | |
96.0 | |
97% | |
2-fluoro-5-chlorosulfonyl benzoic acid, 2-fluoro-5-chlorosulfonylbenzoic acid, 5-chlorosulfonyl-2-fluoro-benzoic acid, 5-chlorosulfonyl-2-fluorobenzoic acid, 5-chlorosulfonyl-2-fluorobenzoicacid, 5-chlorosulphonyl-2-fluorobenzoic acid, acmc-1afaw, benzoic acid, 5-chlorosulfonyl-2-fluoro, ksc577o0r | |
C1=CC(=C(C=C1S(=O)(=O)Cl)C(=O)O)F | |
238.613 | |
238.62 | |
97% |
33.82 to 36.64% C, 1.62 to 1.76% H | |
Glass Bottle | |
111.0°C to 113.0°C | |
37098-75-2 | |
100.0 | |
C7H4ClFO4S | |
CISMZCVQNVNWJW-UHFFFAOYSA-N | |
5-chlorosulfonyl-2-fluorobenzoic acid | |
608984 | |
Crystalline Powder |
Chemical Identifiers
37098-75-2 | |
238.613 | |
2-fluoro-5-chlorosulfonyl benzoic acid, 2-fluoro-5-chlorosulfonylbenzoic acid, 5-chlorosulfonyl-2-fluoro-benzoic acid, 5-chlorosulfonyl-2-fluorobenzoic acid, 5-chlorosulfonyl-2-fluorobenzoicacid, 5-chlorosulphonyl-2-fluorobenzoic acid, acmc-1afaw, benzoic acid, 5-chlorosulfonyl-2-fluoro, ksc577o0r | |
5-chlorosulfonyl-2-fluorobenzoic acid |
C7H4ClFO4S | |
CISMZCVQNVNWJW-UHFFFAOYSA-N | |
608984 | |
C1=CC(=C(C=C1S(=O)(=O)Cl)C(=O)O)F |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger