Please login to your online account to display your discounted pricing

5-Chlorosalicylic Acid 98%, ACROS Organics™

$34.80 - $94.38

Chemical Identifiers

Chemical Name or Material 5-Chlorosalicylic acid
Name Note 98%
CAS 321-14-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H5ClO3
Linear Formula ClC6H3(OH)CO2H
Formula Weight 172.57
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC110071000

  SDS
thermo fisher scientific
110071000
100g Each for $34.80

AC110075000

  SDS
thermo fisher scientific
110075000
500g Each for $94.38
Description & Specifications

Specifications

Chemical Name or Material 5-Chlorosalicylic acid
Color Off-white to cream
Melting Point 171° to 176°C
Quantity 100g
Name Note 98%
CAS 321-14-2
CAS Min % 97.5
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
RTECS Number VO2080000
EINECS Number 206-283-9
Packaging Plastic bottle
TSCA TSCA
Physical Form Powder
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H5ClO3
Linear Formula ClC6H3(OH)CO2H
Formula Weight 172.57
MDL Number MFCD00002457
Beilstein 10, 102
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.