Please login to your online account to display your discounted pricing

5-Chlorosalicylic Acid 98%, ACROS Organics™


Quantity 500g

Manufacturer: thermo fisher scientific  110075000

 View more versions of this product

Catalog No. AC110075000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥97.5%
Formula Weight 172.57
Quantity 500g
Color Off-white to cream
EINECS Number 206-283-9
TSCA TSCA
RTECS Number VO2080000
Melting Point 171° to 176°C
Beilstein 10, 102
CAS Max % 100.0
Chemical Name or Material 5-Chlorosalicylic acid
Assay Percent Range 98%
Molecular Formula C7H5ClO3
Linear Formula ClC6H3(OH)CO2H
MDL Number MFCD00002457
CAS 321-14-2
CAS Min % 97.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Plastic bottle
Name Note 98%
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 5-Chlorosalicylic acid
Name Note 98%
CAS 321-14-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H5ClO3
Linear Formula ClC6H3(OH)CO2H
Formula Weight 172.57
MDL Number MFCD00002457
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents