missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Chlorosalicylanilide, TCI America™

$203.20 - $1,031.27

Chemical Identifiers

CAS 4638-48-6
Molecular Formula C13H10ClNO2
Molecular Weight (g/mol) 247.678
MDL Number MFCD00002320
InChI Key KGYNGVVNFRUOOZ-UHFFFAOYSA-N
PubChem CID 14869
IUPAC Name 5-chloro-2-hydroxy-N-phenylbenzamide
SMILES C1=CC=C(C=C1)NC(=O)C2=C(C=CC(=C2)Cl)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C028625G View Documents TCI America
C028625G
25g Each for $203.20
 
C0286250G View Documents TCI America
C0286250G
250g Each for $1,031.27
 
Specifications

Chemical Identifiers

4638-48-6
247.678
KGYNGVVNFRUOOZ-UHFFFAOYSA-N
5-chloro-2-hydroxy-N-phenylbenzamide
C13H10ClNO2
MFCD00002320
14869
C1=CC=C(C=C1)NC(=O)C2=C(C=CC(=C2)Cl)O

Specifications

White
25g
C13H10ClNO2
KGYNGVVNFRUOOZ-UHFFFAOYSA-N
5-chloro-2-hydroxy-N-phenylbenzamide
14869
Crystal-Powder at 20°C
5-Chlorosalicylanilide
206°C
4638-48-6
MFCD00002320
C1=CC=C(C=C1)NC(=O)C2=C(C=CC(=C2)Cl)O
247.678
247.68
≥98.0% (T)
Safety and Handling

Safety and Handling

RTECSNumber : CV2450475

TSCA : No