missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Chloropyridine-3-boronic acid, 95%, Thermo Scientific™
Chemical Identifiers
CAS | 872041-85-5 |
---|---|
Molecular Formula | C5H5BClNO2 |
Molecular Weight (g/mol) | 157.36 |
InChI Key | NJXYBTMCTZAUEE-UHFFFAOYSA-N |
Synonym | 5-chloropyridine-3-boronic acid, 5-chloropyridin-3-yl boronic acid, 5-chloro-3-pyridineboronic acid, 3-chloropyridine-5-boronic acid, 5-chloro-3-pyridinyl boronic acid, 5-chloro-3-pyridyl boronic acid, 5-chloropyridin-3-yl-3-boronic acid, 3-chloro-5-pyridyl boronic acid, 5-chloro-3-pyridineboronicacid, pubchem17397 |
PubChem CID | 20111877 |
IUPAC Name | (5-chloropyridin-3-yl)boronic acid |
SMILES | B(C1=CC(=CN=C1)Cl)(O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC445360010
|
Thermo Scientific Chemicals
445360010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC445360050
|
Thermo Scientific Chemicals
445360050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
872041-85-5 | |
157.36 | |
5-chloropyridine-3-boronic acid, 5-chloropyridin-3-yl boronic acid, 5-chloro-3-pyridineboronic acid, 3-chloropyridine-5-boronic acid, 5-chloro-3-pyridinyl boronic acid, 5-chloro-3-pyridyl boronic acid, 5-chloropyridin-3-yl-3-boronic acid, 3-chloro-5-pyridyl boronic acid, 5-chloro-3-pyridineboronicacid, pubchem17397 | |
(5-chloropyridin-3-yl)boronic acid |
C5H5BClNO2 | |
NJXYBTMCTZAUEE-UHFFFAOYSA-N | |
20111877 | |
B(C1=CC(=CN=C1)Cl)(O)O |
Specifications
872041-85-5 | |
Glass Bottle | |
1g | |
NJXYBTMCTZAUEE-UHFFFAOYSA-N | |
(5-chloropyridin-3-yl)boronic acid | |
20111877 | |
95% |
94% min. (HPLC) | |
C5H5BClNO2 | |
5-chloropyridine-3-boronic acid, 5-chloropyridin-3-yl boronic acid, 5-chloro-3-pyridineboronic acid, 3-chloropyridine-5-boronic acid, 5-chloro-3-pyridinyl boronic acid, 5-chloro-3-pyridyl boronic acid, 5-chloropyridin-3-yl-3-boronic acid, 3-chloro-5-pyridyl boronic acid, 5-chloro-3-pyridineboronicacid, pubchem17397 | |
B(C1=CC(=CN=C1)Cl)(O)O | |
157.36 | |
157.36 | |
5-Chloropyridine-3-boronic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO â Research Use Only