missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloropentyl acetate, 98%

Manufacturer:  Alfa Aesar™ A1111109

 View more versions of this product

Catalog No. AAA1111109


 

Specifications

Specifications

5-Chloropentyl acetate
96°C to 98°C (11mmHg)
20395-28-2
C7H13ClO2
1752107
ZCYVIAZIVJNAMO-UHFFFAOYSA-N
5-chloropentyl acetate
88521
97°C (206°F)
10g
98%
MFCD00013696
1-Acetoxy-5-chloropentane
CC(=O)OCCCCCCl
164.629
164.63

Chemical Identifiers

20395-28-2
164.629
ZCYVIAZIVJNAMO-UHFFFAOYSA-N
88521
C7H13ClO2
MFCD00013696
1-Acetoxy-5-chloropentane
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents