Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chlorooxindole, 98%

$67.58 - $841.99

Chemical Identifiers

Chemical Name or Material 5-Chlorooxindole
CAS 17630-75-0
Molecular Formula C8H6ClNO
Formula Weight 167.6
MDL Number MFCD00191927
Synonym 5-Chloro-2-indolinone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1921006

 
alfa aesar™
A1921006
5g Each for $67.58

AAA1921014

 
alfa aesar™
A1921014
25g Each for $246.28

AAA1921022

 
alfa aesar™
A1921022
100g Each for $841.99
Description & Specifications

Specifications

Formula Weight 167.6
Chemical Name or Material 5-Chlorooxindole
Assay 98%
Molecular Formula C8H6ClNO
MDL Number MFCD00191927
CAS 17630-75-0
Synonym 5-Chloro-2-indolinone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H361-H302-H317
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P280-P281-P301+P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 196° to 200°C
Quantity 5g

Applications
5-Chloro-2-oxindole was used for the quantitation of 5-chloro-2-oxindole, concomitantly with all of its potential positional isomers using a single, highly specific, normal-phase chromatographic system. 5-Chloro-2-oxindole (5-Chlorooxindole) is a starting material for tenidap sodium, a pharmaceutical drug candidate.

Solubility
Slightly Soluble (2.0 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P281-P301+P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H361-H302-H317
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
May cause an allergic skin reaction.