missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(Chloromethyl)uracil, 97%

Manufacturer:  Alfa Aesar™ 4463906

 View more versions of this product

Catalog No. AA4463906



Specifications

Specifications

5- (Chloromethyl)uracil
3590-48-5
MFCD00218445
UCDUBKRXOPMNGH-UHFFFAOYSA-N
5-(chloromethyl)-1H-pyrimidine-2,4-dione
236125
Powder
5g
C5H5ClN2O2
2,4 1h,3h-pyrimidinedione, 5-chloromethyl
C1=C(C(=O)NC(=O)N1)CCl
160.557
160.56
97%

Chemical Identifiers

3590-48-5
160.557
UCDUBKRXOPMNGH-UHFFFAOYSA-N
236125
C1=C(C(=O)NC(=O)N1)CCl
C5H5ClN2O2
MFCD00218445
2,4 1h,3h-pyrimidinedione, 5-chloromethyl
5-(chloromethyl)-1H-pyrimidine-2,4-dione
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents