missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(Chloromethyl)uracil, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04463901

 View more versions of this product

Catalog No. AA4463901



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5- (Chloromethyl)uracil
C5H5ClN2O2
0.25 g
UCDUBKRXOPMNGH-UHFFFAOYSA-N
5-(chloromethyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione
236125
97%
3590-48-5
MFCD00218445
5-chloromethyl pyrimidine-2,4 1h,3h-dione, 5-chloromethyl-1,3-dihydropyrimidine-2,4-dione, 2,4 1h,3h-pyrimidinedione, 5-chloromethyl, 5-chloromethyluracil, 5-chloromethyl uracil, acmc-1ady4, 5-chloromethyl-2,4-pyrimidinediol, 5-chloromethyl pyrimidine-2,4-diol, 5-chloromethyl uracil 250mg
ClCC1=CNC(=O)NC1=O
160.56
160.56
Powder

Chemical Identifiers

3590-48-5
160.56
UCDUBKRXOPMNGH-UHFFFAOYSA-N
236125
C5H5ClN2O2
MFCD00218445
5-chloromethyl pyrimidine-2,4 1h,3h-dione, 5-chloromethyl-1,3-dihydropyrimidine-2,4-dione, 2,4 1h,3h-pyrimidinedione, 5-chloromethyl, 5-chloromethyluracil, 5-chloromethyl uracil, acmc-1ady4, 5-chloromethyl-2,4-pyrimidinediol, 5-chloromethyl pyrimidine-2,4-diol, 5-chloromethyl uracil 250mg
ClCC1=CNC(=O)NC1=O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only