Please login to your online account to display your discounted pricing

5-(Chloromethyl)-3-(2-thienyl)-1,2,4-oxadiazole, ≥95%, Maybridge

Chemical Identifiers

Chemical Name or Material 5-(Chloromethyl)-3-(2-thienyl)-1, 2, 4-oxadiazole
CAS 63417-81-2
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C7H5ClN2OS
Formula Weight 200.65
MDL Number MFCD03407322
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO00120CB

 
fisher scientific company
MO00120CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 200.65
Quantity 250mg
Assay 95%
CAS Min % 95.0
Assay Percent Range 95%
Molecular Formula C7H5ClN2OS
Chemical Name or Material 5-(Chloromethyl)-3-(2-thienyl)-1, 2, 4-oxadiazole
MDL Number MFCD03407322
CAS 63417-81-2
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Packaging Amber Glass Bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.