missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Chloromethyl-2-oxazolidinone 98%, ACROS Organics™

Manufacturer:  Acros Organics 159290250

 View more versions of this product

Catalog No. AC159290250



Specifications

Specifications

5-Chloromethyl-2-oxazolidinone
Glass bottle
101°C to 105°C
98%
97.5
98%
MFCD00005269
5-chloromethyl-2-oxazolidinone, 5-chloromethyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 5-chloromethyl, 5-chloromethyl oxazolidin-2-one, 5-chloromethyloxazolidin-2-one, 2-oxazolidinone,5-chloromethyl, acmc-209fxq, fnozceqrxkpzez-uhfffaoysa, 5-chloromethyl-2-oxazolidinone, 5-chloromethyl-1,3-oxazolidin-2-one #
C1C(OC(=O)N1)CCl
135.547
135.55
≥97.5%
Authentic
White
25g
22625-57-6
100.0
C4H6ClNO2
27, 145
FNOZCEQRXKPZEZ-UHFFFAOYSA-N
5-(chloromethyl)-1,3-oxazolidin-2-one
90815
Crystalline Powder

Chemical Identifiers

22625-57-6
135.547
FNOZCEQRXKPZEZ-UHFFFAOYSA-N
90815
C1C(OC(=O)N1)CCl
C4H6ClNO2
MFCD00005269
5-chloromethyl-2-oxazolidinone, 5-chloromethyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 5-chloromethyl, 5-chloromethyl oxazolidin-2-one, 5-chloromethyloxazolidin-2-one, 2-oxazolidinone,5-chloromethyl, acmc-209fxq, fnozceqrxkpzez-uhfffaoysa, 5-chloromethyl-2-oxazolidinone, 5-chloromethyl-1,3-oxazolidin-2-one #
5-(chloromethyl)-1,3-oxazolidin-2-one
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 245-137-9

SDS Documents